Learn More
Description
Specifications
Specifications
| Antigen | GOLGA3 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | 0.1M Tris, 0.1M Glycine, and 20% Glycerol (pH 7.0) with 0.01% Thimerosal |
| Gene Accession No. | Q08378 |
| Gene Alias | GCP170Golgi peripheral membrane protein, golgi autoantigen, golgin subfamily a, 3, Golgi complex-associated protein of 170 kD, Golgi complex-associated protein of 170 kDa, Golgi membrane associated protein, golgin A3, Golgin subfamily A member 3, Golgin-160, golgin-165, male enhanced antigen-2, MEA-2, SY2/SY10 protein |
| Gene Symbols | GOLGA3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
