Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLGA5 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB164305 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164305 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164305 Supplier Novus Biologicals Supplier No. NBP281734AF647
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GOLGA5 Polyclonal antibody specifically detects GOLGA5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen GOLGA5
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias Cell proliferation-inducing gene 31 protein, golgi autoantigen, golgin subfamily a, 5, golgin A5, Golgin subfamily A member 5, Golgin-84, GOLIM5, Protein Ret-II, PTC5, RET-fused gene 5 protein, ret-II, RETII, rfg5, RFG5golgi integral membrane protein 5
Host Species Rabbit
Immunogen GOLGA5 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human GOLGA5. The immunogen is located within amino acids 540 - 590 of GOLGA5. Amino Acid Squence: LQSRIKDRDEEIQKLRNQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 9950
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.