Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLGA6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146132 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146132 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146132 Supplier Novus Biologicals Supplier No. NBP146132
Add to Cart
Add to Cart

Goat Polyclonal Antibody

GOLGA6 Polyclonal specifically detects GOLGA6 in Human samples. It is validated for Peptide ELISA.

Specifications

Antigen GOLGA6
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001033729.2
Gene Alias FLJ75859, GLPGolgin-like protein, GOLGA6Golgin linked to PML, golgi autoantigen, golgin subfamily a, 6, golgi autoantigen, golgin subfamily a, 6A, golgi autoantigen, golgin subfamily a, member 6, golgin A6 family, member A, Golgin subfamily A member 6A
Gene Symbols GOLGA6A
Host Species Goat
Immunogen Peptide with sequence C-TLEKADLKTTLYH corresponding to internal region according to NP_001033729.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342096
Test Specificity This antibody recognizes and epitope within the internal region.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.