Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLGA6B Antibody, Novus Biologicals™
SDP

Catalog No. NB397391 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397391 25 μL
NBP233588 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397391 Supplier Novus Biologicals Supplier No. NBP23358825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GOLGA6B Polyclonal specifically detects GOLGA6B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GOLGA6B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. A6NDN3
Gene Alias GOLGA, GOLGA6D, Golgi Autoantigen, Golgin Subfamily A, 6B, Golgin A6 Family, Member B, Golgin Subfamily A Member 6B, Golgin-Like Protein, Putative Golgin Subfamily A Member 6B, Putative Golgin Subfamily A Member 6D
Gene Symbols GOLGA6B
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: ALCAVSTQQQEEDRSSSCREAVLHRRLQQTIKERAL
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55889
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.