Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLGA7B Antibody, Novus Biologicals™
SDP

Catalog No. NBP15675420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15675420 20 μL
NBP156754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15675420 Supplier Novus Biologicals Supplier No. NBP15675420UL

Rabbit Polyclonal Antibody

GOLGA7B Polyclonal specifically detects GOLGA7B in Human samples. It is validated for Western Blot.

Specifications

Antigen GOLGA7B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q2TAP0
Gene Alias bA451M19.3, bA459F3.4, C10orf132, C10orf133, chromosome 10 open reading frame 132, chromosome 10 open reading frame 133, golgi autoantigen, golgin subfamily a, 7B, golgin A7 family, member B, Golgin subfamily A member 7B, MGC131701
Gene Symbols GOLGA7B
Host Species Rabbit
Immunogen Synthetic peptides corresponding to C10ORF132 The peptide sequence was selected from the N terminal of C10ORF132. Peptide sequence MATEVHNLQELRRSASLATKVFIQRDYSDGTICQFQTKFPPELDSRIERQ.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401647
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.