Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLGB1/Giantin Antibody, Novus Biologicals™
SDP

Catalog No. NBP222322 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222322 Supplier Novus Biologicals Supplier No. NBP222322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GOLGB1/Giantin Polyclonal specifically detects GOLGB1/Giantin in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen GOLGB1/Giantin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_004478.3
Gene Alias 372 kDa Golgi complex-associated protein, GCP, GCP372GIANTIN, giantin, golgi autoantigen, golgin subfamily b, macrogolgin (with transmembrane signal)1,macrogolgin, golgi integral membrane protein 1, golgin B1, golgin B1, golgi integral membrane protein, Golgin subfamily B member 1, GOLIM1, Macrogolgin
Gene Symbols GOLGB1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 950 to 981 of human Golgin B1 using the numbering given in entry NP_004478.3 (GeneID 2804).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 2804
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.