Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Golgi Complex Antibody (SPM581), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP234813W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234813W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234813W Supplier Novus Biologicals Supplier No. NBP234813AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Golgi Complex Monoclonal specifically detects Golgi Complex in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Golgi Complex
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM581
Conjugate Alexa Fluor 405
Dilution Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Host Species Mouse
Immunogen SU-DHL-1 large cell lymphoma cells.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This monoclonal antibody recognizes Golgi complex in human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the Golgi complex in human cells only. It can be used to stain the Golgi complex in cell or tissue preparations and can be used as a Golgi marker in subcellular fractions. It produces a diffuse staining pattern of the Golgi zone in normal and malignant cells and may be used to stain Golgi complex of cells in frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations. This monoclonal antibody is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.