Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Golgi Glycoprotein 1/GLG1 Antibody (GLG1/4830), Novus Biologicals™
SDP

Catalog No. NB234650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB234650 100 μg
NB232379 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB234650 Supplier Novus Biologicals Supplier No. NBP320286100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Golgi Glycoprotein 1/GLG1 Monoclonal antibody specifically detects Golgi Glycoprotein 1/GLG1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen Golgi Glycoprotein 1/GLG1
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone GLG1/4830
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 0.5-1 μg/mL, Immunohistochemistry-Frozen 0.5-1 μg/mL
Formulation 10mM PBS with 0.05% BSA
Gene Alias CFR1, CFR-1, cysteine-rich fibroblast growth factor receptor, DKFZp686L08213, E-selectin ligand 1, ESL1, ESL-1, FLJ23319, FLJ23967, FLJ32797, Golgi apparatus protein 1, golgi glycoprotein 1, golgi sialoglycoprotein MG-160, MG160, MG-160
Host Species Mouse
Immunogen Recombinant full-length human Golgi Glycoprotein 1/GLG1 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2734
Target Species Human
Content And Storage Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.