Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Golgicide A
Potent, selective and reversible inhibitor of GBF1 ArfGEF
Specifications
Purity | >98% |
---|---|
Product Type | Golgicide A |
Molecular Weight (g/mol) | 284.3 |
Inhibitors | ARF GTPase Inhibitors |
Description
Potent, specific and reversible inhibitor of Golgi BFA resistance factor 1 (GBF1), an ArfGEF, that decreases Arf1 activation in vivo. Arrests secretion of soluble and membrane-associated proteins at the endoplasmic reticulum-Golgi intermediate compartment. Causes disassembly and dispersal of the Golgi and trans-Golgi network.Specifications
>98% | |
Golgicide A | |
284.3 | |
ARF GTPase Inhibitors |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title