Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Golgicide A
Click to view available options
Quantity:
10 mg
50 mg
Description
Golgicide A is a potent, specific and reversible inhibitor of Golgi BFA resistance factor 1 (GBF1), an ArfGEF, that decreases Arf1 activation in vivo. Arrests secretion of soluble and membrane-associated proteins at the endoplasmic reticulum-Golgi intermediate compartment. Causes disassembly and dispersal of the Golgi and trans-Golgi network.
Specifications
Specifications
Purity | >98% |
Quantity | 50 mg |
Product Type | Golgicide A |
Molecular Weight (g/mol) | 284.3 |
Inhibitors | ARF GTPase Inhibitors |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction