Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Golgin 97 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342533 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342533 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342533 Supplier Novus Biologicals Supplier No. NBP281753MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Golgin 97 Polyclonal antibody specifically detects Golgin 97 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Golgin 97
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias gap junction protein, alpha 4, 37kD, golgi autoantigen, golgin subfamily a, 1, golgin A1, Golgin subfamily A member 1, Golgin-97, MGC33154
Host Species Rabbit
Immunogen Golgin 97 antibody was raised against a 17 amino acid peptide near the amino terminus of human Golgin 97. The immunogen is located within amino acids 30 - 80 of Golgin 97. Amino Acid Squence: DFASDGSSSREDLSSQL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 2800
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.