Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLM1 Antibody (037), Biotin, Novus Biologicals™
SDP

Catalog No. NB037817 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB037817 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB037817 Supplier Novus Biologicals Supplier No. NBP290166B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GOLM1 Monoclonal specifically detects GOLM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GOLM1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 37
Conjugate Biotin
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human GOLM1 (Q8NBJ4-1; Val 40-Leu 401).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51280
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.