Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLM1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP150627 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP150627 0.1 mL
NBP150627SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP150627 Supplier Novus Biologicals Supplier No. NBP150627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GOLM1 Polyclonal specifically detects GOLM1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.

Specifications

Antigen GOLM1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000, Immunocytochemistry/Immunofluorescence 1:500, Knockout Validated
Formulation PBS, 0.1% BSA, and 50% Glycerol with 0.05% Sodium Azide
Gene Accession No. Q8NBJ4
Gene Alias bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257
Gene Symbols GOLM1
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human GOLM1 protein (within residues 250-400). [Swiss-Prot Q8NBJ4]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51280
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.