Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLM1 Antibody (OTI4B12), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP271916C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271916C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271916C Supplier Novus Biologicals Supplier No. NBP271916C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GOLM1 Monoclonal specifically detects GOLM1 in Human, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GOLM1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4B12
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257
Gene Symbols GOLM1
Host Species Mouse
Immunogen Full length recombinant protein of human GOLM1 (NP_808800) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Golgi Apparatus Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51280.0
Target Species Human, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.