Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goosecoid Antibody (OTI1D7), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB272409APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272409APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272409APC Supplier Novus Biologicals Supplier No. NBP272409APC

Mouse Monoclonal Antibody

Goosecoid Monoclonal specifically detects Goosecoid in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Goosecoid
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1D7
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias goosecoid homeobox, homeobox protein goosecoid
Gene Symbols GSC
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 107-257 of human Goosecoid (NP_776248) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 145258.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.