Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goosecoid Antibody (OTI1D7), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB284354 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB284354 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB284354 Supplier Novus Biologicals Supplier No. NBP272409MFV450

Mouse Monoclonal Antibody

Goosecoid Monoclonal antibody specifically detects Goosecoid in Human, Mouse, Rat samples. It is validated for Flow Cytometry

Specifications

Antigen Goosecoid
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1D7
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias goosecoid homeobox, homeobox protein goosecoid
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 107-257 of human GSC (NP_776248) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 145258
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.