Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GORAB Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571029
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1029 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-1029 Supplier Bethyl Laboratories, Inc Supplier No. A305692AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

GORAB encodes a member of the golgin family, a group of coiled-coil proteins localized to the Golgi. The encoded protein may function in the secretory pathway. The encoded protein, which also localizes to the cytoplasm, was identified by interactions with the N-terminal kinase-like protein, and thus it may function in mitosis.

Specifications

Antigen GORAB
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene GORAB
Gene Accession No. Q5T7V8
Gene Alias GO, NTKLBP1, SCYL1BP1, N-terminal kinase-like-binding protein 1, NTKL-binding protein 1, RAB6-interacting golgin, SCY1-like 1 binding protein 1, SCY1-like 1-binding protein 1, SCYL1-BP1, SCYL1-binding protein 1, hNTKL-BP1
Gene Symbols GORAB
Host Species Rabbit
Immunogen Between 325 and 375
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92344
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.