Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GOSR2/Membrin Mouse anti-Human, Clone: 7E7C7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | GOSR2/Membrin |
---|---|
Clone | 7E7C7 |
Concentration | 1.11 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6570
![]() ![]() |
Proteintech Group Inc
661341IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a trafficking membrane protein which transports proteins among the medial- and trans-Golgi compartments. Due to its chromosomal location and trafficking function, this gene may be involved in familial essential hypertension. Three transcript variants encoding three different isoforms have been found for this gene.Specifications
GOSR2/Membrin | |
1.11 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
27 kDa Golgi SNARE protein, Bos1, GOSR2, GS27, Membrin | |
GOSR2 | |
IgG2a | |
Protein A | |
Antibody |
7E7C7 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O14653 | |
9570 | |
GOSR2/Membrin Fusion Protein Ag21393 | |
Primary | |
-20°C | |
GOSR2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title