Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GoTaq™ G2 Colorless Master Mix

GoTaq(R) G2 Master Mixes are premixed ready-to-use solutions containing GoTaq(R) G2 DNA Polymerase, dNTPs, MgCl2 and reaction buffers at optimal concentrations for efficient amplification of a wide range of DNA templates by PCR.

$88.21 - $697.61

Specifications

Content And Storage −30° to −10°C
Description Colorless master mix
For Use With (Application) RT-PCR
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Includes Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Includes Price Quantity & Availability  
PRM7832
View Documents
Promega
M7832
100 reactions Colorless master mix (M783A 2 x 1.25mL), Nuclease-free water (P119A 2 x 1,250μL)
Each for $88.21
Only null left
Add to Cart
 
PRM7833
View Documents
Promega
M7833
1000 reactions Colorless master mix (M783B 1 x 25mL), Nuclease-free water (P119C 1 x 25mL)
Each for $697.61
Only null left
Add to Cart
 
Description

Description

  • All-in-One PCR Master Mix: Just Add Template and Primers
  • High-Performance GoTaq(R) G2 DNA Polymerase in a Ready-to-Use Master Mix
  • Simplifies reaction setup and saves time
  • Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
Specifications

Specifications

−30° to −10°C
Colorless master mix
RT-PCR
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.