Learn More
Description
- Room-Temperature PCR Setup with High-Performance Master Mix
- All-in-one master mix: just add template and primers
- Eliminate nonspecific amplification with a hot start enzyme
- Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
Specifications
Specifications
| Content And Storage | −30° to −10°C |
| No. of Reactions | 100 reactions |
| Sample Type | Colorless master mix |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.