Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GoTaq™ G2 Hot Start Colorless Master Mixes

GoTaq™ G2 Hot Start Master Mixes are ready-to-use mixes containing GoTaq™ G2 Hot Start Polymerase, buffer, dNTPs and optimized magnesium

$176.96 - $1425.17

Specifications

Content And Storage −30° to −10°C
Sample Type Colorless master mix
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
PRM7432
View Documents
Promega
M7432
100 reactions
Each for $176.96
Only null left
Add to Cart
 
PRM7433
View Documents
Promega
M7433
1000 reactions
Each for $1,425.17
Only null left
Add to Cart
 
Description

Description

  • Room-Temperature PCR Setup with High-Performance Master Mix
  • All-in-one master mix: just add template and primers
  • Eliminate nonspecific amplification with a hot start enzyme
  • Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
Specifications

Specifications

−30° to −10°C
Colorless master mix
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.