Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega GoTaq™ G2 Hot Start Polymerase
GoTaq™ G2 Hot Start Polymerase is supplied with 5X Green GoTaq™ Flexi Buffer, 5X Colorless GoTaq™ Flexi Buffer and 25mM MgCl2.
$167.99 - $5722.67
Specifications
Concentration | 5 U/μL |
---|---|
Content And Storage | −30° to −10°C |
Polymerase | GoTaq G2 |
Sample Type | Hot Start Polymerase |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PRM7401
|
Promega
M7401 |
100U |
Each for $167.99
|
|
|||||
PRM7405
|
Promega
M7405 |
500U |
Each for $400.08
|
|
|||||
PRM7406
|
Promega
M7406 |
2500U |
Each for $1,753.09
|
|
|||||
PRM7408
|
Promega
M7408 |
10,000U |
Each for $5,722.67
|
|
|||||
Description
- High Performance GoTaq(R) G2 Enzyme with Convenient Room-Temperature Setup
- Eliminate nonspecific amplification with a hot start enzyme
- Detect lower-copy-number targets
- Optimize reaction conditions with a Flexi Reaction Buffer and separate MgCl2
- Supplied at a concentration of 5u/μL
Specifications
5 U/μL | |
−30° to −10°C | |
GoTaq G2 | |
Hot Start Polymerase |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title