Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP-IX Antibody (GRP-P), Novus Biologicals™
SDP

Catalog No. NB10065904 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065904 0.1 mL
NB017754 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10065904 Supplier Novus Biologicals Supplier No. NB10065904
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GP-IX Monoclonal specifically detects GP-IX in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen GP-IX
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone GRP-P
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:100, Immunoprecipitation 1:10-1:500
Gene Accession No. P14770
Gene Alias CD42a, CD42a antigen, Glycoprotein 9, glycoprotein IX (platelet), GP-IX, GPIXplatelet glycoprotein IX
Gene Symbols GP9
Host Species Mouse
Immunogen Human red blood cells and platelets
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2815
Test Specificity Recognizes the platelet GP-IX antigen, a 23kD surface marker expressed by platelets and magakaryocytes. The GP-IX complex is the major platelet receptor for von Willebrand factor.
Target Species Human, Canine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.