Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gp130/CD130 Antibody (125623), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347645 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347645 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347645 Supplier Novus Biologicals Supplier No. FAB4681MFV610
Add to Cart
Add to Cart

Rat Monoclonal Antibody

gp130/CD130 Monoclonal antibody specifically detects gp130/CD130 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen gp130/CD130
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 125623
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse gp130, Gln23-Glu617 (predicted), Accession # Q6PDI9
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3572
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.