Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gp130/CD130 Antibody (745314), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB312167 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312167 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312167 Supplier Novus Biologicals Supplier No. FAB5029PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

gp130/CD130 Monoclonal antibody specifically detects gp130/CD130 in Rat samples. It is validated for Flow Cytometry

Specifications

Antigen gp130/CD130
Applications Flow Cytometry
Classification Monoclonal
Clone 745314
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat gp130, Gln23-Glu618 (predicted), Accession # P40190
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3572
Target Species Rat
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.