Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ gp130/CD130 Antibody Pair [HRP]
SDP

Catalog No. NB010110 Shop All Novus Biologicals Products
Click to view available options
Quantity:
15 Plates
5 Plates

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse gp130/CD130.

Specifications

Assay Range 11.72 pg/mL
Assay Sensitivity 11.72-750 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse gp130/CD130.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
Gene ID (Entrez) 3572
Gene Symbol IL6ST
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.2 mg/mL rabbit anti-gp130/CD130 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-mouse gp130/CD130 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 60 ng of recombinant mouse gp130/CD130.
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Cancer, Immunology
    Storage Requirements Storage is content dependent.
    Target gp130/CD130
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use.. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 750 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.