Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP1BB Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB031980 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB031980 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB031980 Supplier Novus Biologicals Supplier No. NBP2840520.1ML

Rabbit Polyclonal Antibody

GP1BB Polyclonal specifically detects GP1BB in Human samples. It is validated for Western Blot.

Specifications

Antigen GP1BB
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias antigen CD42b-beta, BDPLT1, BS, glycoprotein Ib (platelet), beta polypeptide, glycoprotein Ib platelet beta subunit, GPIBB, GPIbbeta, GPIb-beta, nuclear localization signal deleted in velocardiofacial syndrome, platelet glycoprotein Ib beta chain, platelet membrane glycoprotein Ib beta, truncated platelet membrane glycoprotein Ib beta
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C terminal region of human GP1BB
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2812
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.