Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP2 Antibody (GP2/1805) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB118941 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB118941 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB118941 Supplier Novus Biologicals Supplier No. NBP308215
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GP2 Monoclonal specifically detects GP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen GP2
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone GP2/1805
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-2 ug/mL, ELISA : 2-4 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75
Host Species Mouse
Immunogen Recombinant fragment of human GP2 protein (around aa 35-179) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2813
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.