Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP2 Antibody (GP2/2569R), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB382481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382481 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382481 Supplier Novus Biologicals Supplier No. NBP308242AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GP2 Monoclonal antibody specifically detects GP2 in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen GP2
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GP2/2569R
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75
Host Species Rabbit
Immunogen Recombinant human GP2 protein fragment (around aa35-179) (exact sequence is proprietary) (Uniprot: P55259)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2813
Target Species Human, Mouse (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.