Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gp96/HSP90B1/GRP94 Antibody (2H3), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP104346FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104346FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104346FR Supplier Novus Biologicals Supplier No. NBP104346FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

gp96/HSP90B1/GRP94 Monoclonal specifically detects gp96/HSP90B1/GRP94 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen gp96/HSP90B1/GRP94
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2H3
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Gene Accession No. NP_003290
Gene Alias 94 kDa glucose-regulated protein, endothelial cell (HBMEC) glycoprotein, glucose regulated protein, 94 kDa, GP96ECGP, GRP-94, GRP94gp96 homolog, Heat shock protein 90 kDa beta member 1, heat shock protein 90kDa beta (Grp94), member 1, TRA1endoplasmin, tumor rejection antigen (gp96) 1, Tumor rejection antigen 1, Tumor rejection antigen-1 (gp96)
Gene Symbols HSP90B1
Host Species Mouse
Immunogen Recombinant human TRA1 (676-803aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 7184
Test Specificity Anti-human TRA1 mAb, clone 2H3, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TRA1 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.