Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gp96/HSP90B1/GRP94 Antibody (9G10.F8.2), Novus Biologicals™
SDP

Catalog No. NBP24468902 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24468902 0.2 mg
NBP24468900 0.02 mg
NBP24468901 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP24468902 Supplier Novus Biologicals Supplier No. NBP2446890.2MG

Rat Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

gp96/HSP90B1/GRP94 Monoclonal antibody specifically detects gp96/HSP90B1/GRP94 in Human, Mouse, Rat, Porcine, Canine, Chicken, Equine, Guinea Pig, Hamster, Monkey, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen gp96/HSP90B1/GRP94
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone 9G10.F8.2
Concentration 0.2mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias 94 kDa glucose-regulated protein, endothelial cell (HBMEC) glycoprotein, glucose regulated protein, 94 kDa, GP96ECGP, GRP-94, GRP94gp96 homolog, Heat shock protein 90 kDa beta member 1, heat shock protein 90kDa beta (Grp94), member 1, TRA1endoplasmin, tumor rejection antigen (gp96) 1, Tumor rejection antigen 1, Tumor rejection antigen-1 (gp96)
Gene Symbols HSP90B1
Host Species Rat
Immunogen Purified glucose regulated protein 94 (grp94) from chicken oviducts
Molecular Weight of Antigen 94 kDa
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 7184
Test Specificity Recognizes a protein of 94kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This MAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grp s are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.
Target Species Human, Mouse, Rat, Pig, Canine, Chicken, Equine, Guinea Pig, Hamster, Monkey, Primate, Rabbit, Sheep, Xenopus species
Content And Storage Store at 4C.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.