Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gp96/HSP90B1/GRP94 Antibody (HSP90B1/3168R), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB384604 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384604 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384604 Supplier Novus Biologicals Supplier No. NBP308763AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

gp96/HSP90B1/GRP94 Monoclonal antibody specifically detects gp96/HSP90B1/GRP94 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Equine, Guinea Pig, Hamster, Monkey, Rabbit, Sheep, Xenopus samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen gp96/HSP90B1/GRP94
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HSP90B1/3168R
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias 94 kDa glucose-regulated protein, endothelial cell (HBMEC) glycoprotein, glucose regulated protein, 94 kDa, GP96ECGP, GRP-94, GRP94gp96 homolog, Heat shock protein 90 kDa beta member 1, heat shock protein 90kDa beta (Grp94), member 1, TRA1endoplasmin, tumor rejection antigen (gp96) 1, Tumor rejection antigen 1, Tumor rejection antigen-1 (gp96)
Host Species Rabbit
Immunogen Recombinant full-length human gp96/HSP90B1/GRP94 protein (Uniprot: P14625)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cellular Markers, Cytokine Research, ER Markers, Golgi Apparatus Markers, Hypoxia, Immunology, Membrane Trafficking and Chaperones, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7184
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.