Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPAA1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096354 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096354 20 μL
NB096353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096354 Supplier Novus Biologicals Supplier No. NBP30518120UL

Rabbit Polyclonal Antibody

GPAA1 Polyclonal specifically detects GPAA1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen GPAA1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias anchor attachment protein 1 (Gaa1p, yeast) homolog, GAA1 protein homolog, GAA1glycophosphatidylinositol anchor attachment 1, glycosylphosphatidylinositol anchor attachment protein 1 homolog (yeast), GPAA1P anchor attachment protein 1 homolog, GPAA1P anchor attachment protein 1 homolog (yeast), GPI anchor attachment protein 1, GPI transamidase subunit, hGAA1glycosylphosphatidylinositol anchor attachment 1 protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 90-310 of human GPAA1 (NP_003792.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8733
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.