Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPER/GPR30 Antibody, Novus Biologicals™
SDP

Catalog No. NBP131239 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131239 Supplier Novus Biologicals Supplier No. NBP131239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

GPER/GPR30 Polyclonal specifically detects GPER/GPR30 in Human, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen GPER/GPR30
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q99527
Gene Alias CEPRG-protein coupled receptor 30, Chemoattractant receptor-like 2, chemokine receptor-like 2, CMKRL2MGC99678, constitutively expressed peptide-like receptor, DRY12IL8-related receptor DRY12, FEG-1mER, Flow-induced endothelial G-protein coupled receptor 1, G protein-coupled estrogen receptor 1, G protein-coupled receptor 30, GPCR-BR, GPR30GPCR-Br, G-protein coupled estrogen receptor 1, heptahelix receptor, LERGU, LERGU2, leucine rich protein in GPR30 3'UTR, LyGPR, Lymphocyte-derived G-protein coupled receptor, Membrane estrogen receptor
Gene Symbols GPER1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GPR30. The exact sequence is proprietary.
Molecular Weight of Antigen 42 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2852
Target Species Human, Pig
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.