Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPHA2 Antibody (06), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB353096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB353096 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB353096 Supplier Novus Biologicals Supplier No. NBP306201MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPHA2 Monoclonal antibody specifically detects GPHA2 in Human samples. It is validated for ELISA

Specifications

Antigen GPHA2
Applications ELISA
Classification Monoclonal
Clone 06
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias A2, glycoprotein alpha 2, glycoprotein hormone alpha 2, glycoprotein hormone alpha-2, GPA2Thyrostimulin subunit alpha, MGC126572, Putative secreted protein Zsig51, ZSIG51cysteine knot protein
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GPHA2 (Uniprot#: Q96T91; Met1-Tyr129).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170589
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.