Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPHA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140034
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5140034 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIPA5140034

Supplier: Invitrogen™ PA5140034

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GQEAVIPGCH LHPFNVTVRS DRQGTCQGSH VAQAC Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 86%.

GPHA2 is a cystine knot-forming polypeptide and a subunit of the dimeric glycoprotein hormone family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPHA2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gpha2
Gene Accession No. Q96T91
Gene Alias A2; cysteine knot protein; glycoprotein alpha 2; glycoprotein hormone alpha 2; glycoprotein hormone alpha-2; Gpa2; Gpha2; Putative secreted protein Zsig51; thyrostimulin subunit alpha; ZSIG51
Gene Symbols Gpha2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPHA2. Recombinant protein control fragment (Product #RP-109121).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170589
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.