Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPHN Monoclonal Antibody (OTI5C9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502313S


Description
GPHN Monoclonal antibody specifically detects GPHN in Human, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
GPHN | |
Monoclonal | |
0.76 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q03555, Q9NQX3 | |
Mouse | |
Affinity chromatography | |
RUO | |
10243, 64845 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Western Blot | |
OTI5C9 | |
Unconjugated | |
Gphn | |
5730552E08Rik; AI662856; BC027112; C230040D23; Domain E; Domain G; EGK_18299; Geph; gephyrin; gephyrin isoform; Gph; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC; Molybdopterin adenylyltransferase; Molybdopterin molybdenumtransferase; MPT adenylyltransferase; MPT Mo-transferase; Putative glycine receptor-tubulin linker protein | |
Full length human recombinant protein of human GPHN produced in HEK293T cell. | |
30 μL | |
Primary | |
Human, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction