Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPHN Monoclonal Antibody (OTI5C9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
$231.00 - $516.29
Specifications
Antigen | GPHN |
---|---|
Clone | OTI5C9 |
Concentration | 0.76 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-254-1642
![]() ![]() ![]() |
OriGene
TA502313S |
30 μL |
Each for $231.00
|
|
|||||
50-167-3376
![]() ![]() |
OriGene
TA502313 |
100 μL |
Each for $516.29
|
|
|||||
Description
GPHN Monoclonal antibody specifically detects GPHN in Human, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
GPHN | |
0.76 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Primary |
OTI5C9 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Mouse | |
IgG2a | |
-20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title