Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPI-PLD/GPLD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB092593 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092593 100 μL
NB092594 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092593 Supplier Novus Biologicals Supplier No. NBP303599100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPI-PLD/GPLD1 Polyclonal specifically detects GPI-PLD/GPLD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GPI-PLD/GPLD1
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EC 3.1.4.50, Glycoprotein phospholipase D, glycosylphosphatidylinositol specific phospholipase D1, glycosylphosphatIdylinositol-specific phospholipase D, Glycosyl-phosphatidylinositol-specific phospholipase D, GPIPLD, GPI-PLD, GPIPLDM, GPI-specific phospholipase D, MGC22590, phosphatidylinositol-glycan-specific phospholipase D, phospholipase D, phosphatidylinositol-glycan-specific, PI-G PLD, PIGPLD, PIGPLD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-160 of human GPI-PLD/GPLD1 (NP_001494.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2822
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.