Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPI-PLD/GPLD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GPI-PLD/GPLD1 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GPI-PLD/GPLD1 Polyclonal specifically detects GPI-PLD/GPLD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GPI-PLD/GPLD1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
2822 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EC 3.1.4.50, Glycoprotein phospholipase D, glycosylphosphatidylinositol specific phospholipase D1, glycosylphosphatIdylinositol-specific phospholipase D, Glycosyl-phosphatidylinositol-specific phospholipase D, GPIPLD, GPI-PLD, GPIPLDM, GPI-specific phospholipase D, MGC22590, phosphatidylinositol-glycan-specific phospholipase D, phospholipase D, phosphatidylinositol-glycan-specific, PI-G PLD, PIGPLD, PIGPLD1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 24-160 of human GPI-PLD/GPLD1 (NP_001494.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title