Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPIHBP1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB339322 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339322 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339322 Supplier Novus Biologicals Supplier No. NB11041539APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPIHBP1 Polyclonal antibody specifically detects GPIHBP1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen GPIHBP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to an internal region [within residues 170-250] of mouse GPIHBP1. [Swiss-Prot# Q9D1N2]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 338328
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.