Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPIHBP1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB080806 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB080806 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB080806 Supplier Novus Biologicals Supplier No. NBP297041H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPIHBP1 Polyclonal specifically detects GPIHBP1 in Human samples. It is validated for ELISA.

Specifications

Antigen GPIHBP1
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant GPIHBP1 (AAH63857.1; Met1-Ser160).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 338328
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.