Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPIHBP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB11055293T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11055293T 0.025 mL
NB11055293 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11055293T Supplier Novus Biologicals Supplier No. NB11055293SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPIHBP1 Polyclonal specifically detects GPIHBP1 in Human samples. It is validated for Western Blot.

Specifications

Antigen GPIHBP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml
Formulation Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
Gene Accession No. Q8IV16
Gene Alias glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1
Gene Symbols GPIHBP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to an internal region [within residues 100-180] of human GPIHBP1. [Swiss-Prot# Q8IV16]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 338328
Test Specificity Human. Immunogen has 78% homology to mouse.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.