Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPIHBP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | GPIHBP1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 2 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11055293T
![]() |
Novus Biologicals
NB11055293SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB11055293
![]() |
Novus Biologicals
NB11055293 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
GPIHBP1 Polyclonal specifically detects GPIHBP1 in Human samples. It is validated for Western Blot.Specifications
GPIHBP1 | |
Western Blot 2 ug/ml | |
Polyclonal | |
Rabbit | |
Cholesterol Metabolism, Lipid and Metabolism | |
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1 | |
GPIHBP1 | |
IgG | |
Affinity Purified | |
Human. Immunogen has 78% homology to mouse. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Q8IV16 | |
338328 | |
A synthetic peptide corresponding to an internal region [within residues 100-180] of human GPIHBP1. [Swiss-Prot# Q8IV16] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title