Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPN2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15535920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15535920 20 μL
NBP155359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15535920 Supplier Novus Biologicals Supplier No. NBP15535920UL

Rabbit Polyclonal Antibody

GPN2 Polyclonal specifically detects GPN2 in Human samples. It is validated for Western Blot.

Specifications

Antigen GPN2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9H9Y4
Gene Alias ATP binding domain 1 family, member B, ATPBD1B, ATP-binding domain 1 family member B, FLJ10349, GPN-loop GTPase 2
Gene Symbols GPN2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to GPN2 (GPN-loop GTPase 2) The peptide sequence was selected from the middle region of GPN2. Peptide sequence VLQAVDKANGYCFRAQEQRSLEAMMSAAMGADFHFSSTLGIQEKYLAPSN.
Molecular Weight of Antigen 34 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54707
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.