Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPN3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB350472 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350472 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350472 Supplier Novus Biologicals Supplier No. NBP318861MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPN3 Polyclonal antibody specifically detects GPN3 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen GPN3
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ATP binding domain 1 family, member C, ATPBD1C, ATP-binding domain 1 family member C, GPN-loop GTPase 3, MGC14560, MGC32810, protein x 0004
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GPN3 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51184
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.