Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR101 Antibody (909603) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB374523 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374523 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374523 Supplier Novus Biologicals Supplier No. FAB7495J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR101 Monoclonal specifically detects GPR101 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen GPR101
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 909603
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor 101, GPCR6, probable G-protein coupled receptor 101
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR101, Met1-Pro508,, Accession # NP_473362
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 83550
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.