Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR109B/HM74 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224488 0.1 mg
NBP224488SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224488 Supplier Novus Biologicals Supplier No. NBP224488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR109B/HM74 Polyclonal specifically detects GPR109B/HM74 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GPR109B/HM74
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
Gene Alias G protein-coupled receptor 109B, G-protein coupled receptor 109B, G-protein coupled receptor HM74, G-protein coupled receptor HM74B, GTP-binding protein, HCA3, HCAR3, HM74B, HM74Puma-g, Niacin receptor 2, NIACR2, Nicotinic acid receptor 2, PUMAG, putative chemokine receptor
Gene Symbols HCAR3
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human GPR109B was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, GPCR, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8843
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 86% in rhesus monkey and 73% homologous in bovine and human REG3G.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.