Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR111 Antibody (594522), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB346978 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346978 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346978 Supplier Novus Biologicals Supplier No. FAB64941MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR111 Monoclonal specifically detects GPR111 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen GPR111
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 594522
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor 111
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human GPR111, Cys19-Lys375, Accession # Q8IZF7
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 222611
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.