Learn More
Description
Specifications
Specifications
| Antigen | GPR146 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | G protein-coupled receptor 146, G protein-coupled receptor PGR8, PGR8G-protein coupled receptor PGR8, probable G-protein coupled receptor 146 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle terminal region of mouse GPR146 (NP_001033792.1). Peptide sequence ERALPRTYMASVYNTRHVCGFVWGGAVLTSFSSLLFYICSHVSSRIAECA |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
